Assay Details
Target Gene Details
Entrez Gene ID: | 67041 |
Gene Name: | 3-oxoacid CoA transferase 1 |
Gene Aliases: |
2610008O03Rik, Oxct, Oxct2a, Scot-s |
Location: |
Chr.15:4026428-4155344 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oxct1 | NM_024188.6 | NP_077150.1 | ||
| AB085609.1 | BAC05524.1 | |||
| AK009027.1 | BAB26035.1 | |||
| AK010790.1 | BAB27184.3 | |||
| AK011354.1 | BAB27562.1 | |||
| AK078086.1 | ||||
| AK146158.1 | BAE26941.1 | |||
| AK146342.1 | BAE27096.1 | |||
| AK151662.1 | BAE30590.1 | |||
| AK151695.1 | BAE30619.1 | |||
| AK167681.1 | BAE39730.1 | |||
| AK181182.1 | ||||
| AK181314.1 | ||||
| AK184597.1 | ||||
| AK185295.1 | ||||
| BC003422.1 | AAH03422.1 | |||
| EF095212.1 |
More Information
Set Membership: |
|

Genomic Map