Assay Details
Target Gene Details
Entrez Gene ID: | 67041 |
Gene Name: | 3-oxoacid CoA transferase 1 |
Gene Aliases: |
2610008O03Rik, Oxct, Oxct2a, Scot-s |
Location: |
Chr.15:4026428-4155344 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oxct1 | NM_024188.6 | 8 | 1120 | NP_077150.1 |
| AB085609.1 | 8 | 862 | BAC05524.1 | |
| AK009027.1 | 8 | 900 | BAB26035.1 | |
| AK010790.1 | 8 | 863 | BAB27184.3 | |
| AK011354.1 | 8 | 800 | BAB27562.1 | |
| AK078086.1 | 8 | 1120 | ||
| AK146158.1 | 8 | 878 | BAE26941.1 | |
| AK146342.1 | 8 | 865 | BAE27096.1 | |
| AK151662.1 | 8 | 859 | BAE30590.1 | |
| AK151695.1 | 8 | 850 | BAE30619.1 | |
| AK167681.1 | 8 | 950 | BAE39730.1 | |
| AK181182.1 | ||||
| AK185295.1 | ||||
| AK209817.1 | ||||
| AK209998.1 | ||||
| AK215681.1 | ||||
| BC003422.1 | 8 | 824 | AAH03422.1 |
More Information
Set Membership: |
|

Genomic Map