Assay Details
Target Gene Details
Entrez Gene ID: | 19082 |
Gene Name: | protein kinase, AMP-activated, gamma 1 non-catalytic subunit |
Gene Aliases: |
AA571379, BB036179, Prkaac |
Location: |
Chr.15:98812797-98831508 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkag1 | NM_016781.2 | NP_058061.2 | ||
| AF036535.1 | AAB95475.1 | |||
| AK138324.1 | BAE23624.1 | |||
| AK159583.1 | BAE35203.1 | |||
| BC086660.1 | AAH86660.1 | |||
| BC109015.1 | AAI09016.1 | |||
| BQ963077.1 |
More Information
Set Membership: |
|

Genomic Map