Assay Details
Target Gene Details
Entrez Gene ID: | 110310 |
Gene Name: | keratin 7 |
Gene Aliases: |
D15Wsu77e, K7, Krt2-7 |
Location: |
Chr.15:101412403-101427806 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt7 | NM_033073.3 | 7 | 1097 | NP_149064.1 |
| AF509888.1 | 7 | 1074 | AAN64032.1 | |
| AK002434.1 | 7 | 1075 | BAB22099.1 | |
| AK146914.1 | 7 | 1075 | BAE27526.1 | |
| BC030403.1 | 7 | 1108 | AAH30403.1 |

Genomic Map