Assay Details
Target Gene Details
Entrez Gene ID: | 110310 |
Gene Name: | keratin 7 |
Gene Aliases: |
D15Wsu77e, K7, Krt2-7 |
Location: |
Chr.15:101412403-101427806 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt7 | NM_033073.3 | NP_149064.1 | ||
| AF509888.1 | AAN64032.1 | |||
| AK002434.1 | BAB22099.1 | |||
| AK146914.1 | BAE27526.1 | |||
| BC030403.1 | AAH30403.1 | |||
| BU755120.1 |
More Information
Set Membership: |
|

Genomic Map