Assay Details
Target Gene Details
Entrez Gene ID: | 114566 |
Gene Name: | keratin 82 |
Gene Aliases: |
Krt2-20 |
Location: |
Chr.15:101541216-101550659 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt82 | NM_053249.3 | NP_444479.2 | ||
| AY028606.1 | AAK26232.1 | |||
| BC108971.2 | AAI08972.1 |
More Information
Set Membership: |
|

Genomic Map