Assay Details
Target Gene Details
Entrez Gene ID: | 105847 |
Gene Name: | lipase maturation factor 2 |
Gene Aliases: |
AI451006, Tmem112b, Tmem153 |
Location: |
Chr.15:89351004-89355659 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lmf2 | NM_178919.4 | NP_849250.1 | ||
| AK042427.1 | ||||
| AK084606.1 | BAC39228.1 | |||
| AK085435.1 | ||||
| AK149916.1 | BAE29164.1 | |||
| AK149934.1 | BAE29177.1 | |||
| AK158371.1 | ||||
| AK171488.1 | BAE42488.1 | |||
| BC079532.1 | AAH79532.1 |

Genomic Map