Assay Details
Target Gene Details
Entrez Gene ID: | 170719 |
Gene Name: | oxidation resistance 1 |
Gene Aliases: |
2210416C20Rik, C7, C7B |
Location: |
Chr.15:41447482-41861048 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oxr1 | NM_001130165.1 | 1 | 59 | NP_001123637.1 |
| NM_001130166.1 | NP_001123638.1 | |||
| AK052714.1 | BAC35112.2 | |||
| AK158703.1 | BAE34619.1 | |||
| BC089183.1 | 1 | 54 | AAH89183.1 | |
| BC098491.1 | AAH98491.1 | |||
| CJ178211.1 | 1 | 154 |

Genomic Map