Assay Details
Target Gene Details
Entrez Gene ID: | 22293 |
Gene Name: | solute carrier family 45, member 2 |
Gene Aliases: |
Aim-1, Aim1, Dbr, Matp, Oca4, blanc-sale, bls, uw |
Location: |
Chr.15:10995693-11029233 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc45a2 | NM_053077.3 | NP_444307.1 | ||
| AF360357.1 | AAK81713.1 | |||
| AK029155.1 | BAC26330.1 | |||
| AK089932.1 | BAC41003.1 | |||
| AK211368.1 | ||||
| AY034377.1 | AAK58462.1 | |||
| BC125376.1 | ||||
| BC132429.1 |
More Information
Set Membership: |
|

Genomic Map