Assay Details
Target Gene Details
Entrez Gene ID: | 406219 |
Gene Name: | keratin 83 |
Gene Aliases: |
Krt2-25, Krt87 |
Location: |
Chr.15:101431490-101438804 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt83 | NM_001003668.2 | NP_001003668.2 | ||
| BC139321.1 | ||||
| BC139322.1 |

Genomic Map