Assay Details
Target Gene Details
Entrez Gene ID: | 239393 |
Gene Name: | low density lipoprotein-related protein 12 |
Gene Aliases: |
AI848829, C820005L12Rik, LRP-12 |
Location: |
Chr.15:39870602-39944009 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrp12 | NM_172814.3 | NP_766402.1 | ||
| AK050512.1 | BAC34299.1 | |||
| BC058345.1 | AAH58345.1 |
More Information
Set Membership: |
|

Genomic Map