Assay Details
Target Gene Details
Entrez Gene ID: | 170719 |
Gene Name: | oxidation resistance 1 |
Gene Aliases: |
2210416C20Rik, C7, C7B |
Location: |
Chr.15:41447482-41861048 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oxr1 | NM_001130163.1 | NP_001123635.1 | ||
| NM_001130164.1 | NP_001123636.1 | |||
| NM_001130165.1 | NP_001123637.1 | |||
| NM_001130166.1 | NP_001123638.1 | |||
| NM_130885.2 | NP_570955.1 | |||
| AF324899.1 | AAK30368.1 | |||
| AF333985.1 | AAK29400.1 | |||
| AK158703.1 | BAE34619.1 | |||
| AW106090.1 | ||||
| BC003927.1 | AAH03927.1 | |||
| BC089183.1 | AAH89183.1 | |||
| BC098491.1 | AAH98491.1 |

Genomic Map