Assay Details
Target Gene Details
Entrez Gene ID: | 20401 |
Gene Name: | SH3-domain binding protein 1 |
Gene Aliases: |
3BP-1 |
Location: |
Chr.15:78899780-78912052 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sh3bp1 | NM_009164.2 | NP_033190.2 | ||
| AK145156.1 | ||||
| AK161215.1 | ||||
| AK166337.1 | ||||
| AK172060.1 | ||||
| BC004598.1 | AAH04598.1 | |||
| X87671.1 | CAA61011.1 |

Genomic Map