Assay Details
Target Gene Details
Entrez Gene ID: | 268783 |
Gene Name: | myotubularin related protein 12 |
Gene Aliases: |
3Pap, 4932703C11, C730015A02Rik, Pip3ap, mKIAA1682 |
Location: |
Chr.15:12205094-12272240 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtmr12 | NM_172958.3 | NP_766546.1 | ||
| AK030139.1 | BAC26803.1 | |||
| AK050099.1 | ||||
| AK050236.1 | ||||
| AK076161.1 | BAC36227.1 | |||
| AK122539.1 | BAC65821.1 | |||
| BC054352.1 | AAH54352.1 |
More Information
Set Membership: |
|

Genomic Map