Assay Details
Target Gene Details
Entrez Gene ID: | 170719 |
Gene Name: | oxidation resistance 1 |
Gene Aliases: |
2210416C20Rik, C7, C7B |
Location: |
Chr.15:41447482-41861048 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oxr1 | NM_001130163.1 | 3 | 245 | NP_001123635.1 |
| NM_001130165.1 | 3 | 340 | NP_001123637.1 | |
| NM_001130166.1 | 5 | 604 | NP_001123638.1 | |
| NM_130885.2 | 3 | 245 | NP_570955.1 | |
| AF324899.1 | 3 | 244 | AAK30368.1 | |
| AF333985.1 | 3 | 245 | AAK29400.1 | |
| AK040881.1 | 3 | 267 | ||
| AK052714.1 | 3 | 410 | BAC35112.2 | |
| AK158703.1 | 5 | 586 | BAE34619.1 | |
| BC089183.1 | 3 | 335 | AAH89183.1 | |
| BC098491.1 | 5 | 587 | AAH98491.1 | |
| CJ178211.1 |

Genomic Map