Assay Details
Target Gene Details
Entrez Gene ID: | 170719 |
Gene Name: | oxidation resistance 1 |
Gene Aliases: |
2210416C20Rik, C7, C7B |
Location: |
Chr.15:41447482-41861048 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oxr1 | NM_001130163.1 | 8 | 1576 | NP_001123635.1 |
| NM_001130165.1 | 8 | 1671 | NP_001123637.1 | |
| NM_001130166.1 | 10 | 1935 | NP_001123638.1 | |
| NM_130885.2 | 8 | 1576 | NP_570955.1 | |
| AF324899.1 | 8 | 1575 | AAK30368.1 | |
| AF333985.1 | 8 | 1576 | AAK29400.1 | |
| AK040881.1 | 8 | 1599 | ||
| AK158703.1 | 10 | 1917 | BAE34619.1 | |
| BC003927.1 | 2 | 166 | AAH03927.1 | |
| BC089183.1 | 8 | 1666 | AAH89183.1 | |
| BC098491.1 | 10 | 1918 | AAH98491.1 |

Genomic Map