Assay Details
Target Gene Details
Entrez Gene ID: | 58234 |
Gene Name: | SH3/ankyrin domain gene 3 |
Gene Aliases: |
AI841104, SHANK3c-3, SHANK3c-4, Shank3b |
Location: |
Chr.15:89499707-89560261 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Shank3 | NM_021423.3 | NP_067398.2 | ||
| AB231013.1 | BAE16756.1 | |||
| AJ245904.1 | CAB89816.1 | |||
| KF612592.1 |

Genomic Map