Assay Details
Target Gene Details
Entrez Gene ID: | 69790 |
Gene Name: | mediator complex subunit 30 |
Gene Aliases: |
1810038N03Rik, 2510044J04Rik, TRAP25, Thrap6 |
Location: |
Chr.15:52712445-52730431 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med30 | NM_027212.2 | 1 | 202 | NP_081488.1 |
| AK007739.1 | 1 | 194 | BAB25226.1 | |
| AK011104.1 | 1 | 203 | BAB27401.1 | |
| AK146250.1 | 1 | 206 | BAE27012.1 | |
| BC020113.1 | 1 | 189 | AAH20113.1 |
More Information
Set Membership: |
|

Genomic Map