Assay Details
Target Gene Details
Entrez Gene ID: | 68479 |
Gene Name: | PHD finger protein 5A |
Gene Aliases: |
1110007B08Rik |
Location: |
Chr.15:81864516-81871892 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phf5a | NM_026737.3 | NP_081013.1 | ||
| AF479288.1 | AAM49736.1 | |||
| AK003520.1 | BAB22833.1 | |||
| AK143590.1 | BAE25453.1 | |||
| AK163247.1 | BAE37255.1 | |||
| AK167932.1 | BAE39936.1 | |||
| AK167955.1 | BAE39955.1 | |||
| BC025161.1 | AAH25161.1 |

Genomic Map