Assay Details
Target Gene Details
Entrez Gene ID: | 14083 |
Gene Name: | PTK2 protein tyrosine kinase 2 |
Gene Aliases: |
FAK, FRNK, Fadk, mKIAA4203, p125FAK |
Location: |
Chr.15:73205102-73423275 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptk2 | NM_001130409.1 | 23 | 2357 | NP_001123881.1 |
| NM_007982.2 | 22 | 2202 | NP_032008.2 | |
| AB011499.1 | 23 | 2013 | BAC53890.1 | |
| AB030035.1 | BAC53924.1 | |||
| AK079821.1 | 23 | 2454 | BAC37757.1 | |
| AK220543.1 | 22 | 2202 | BAD90317.1 | |
| JN971016.1 | 22 | 1929 | ||
| M95408.1 | 21 | 1999 | AAA37592.1 |

Genomic Map