Assay Details
Target Gene Details
Entrez Gene ID: | 21357 |
Gene Name: | TAR (HIV) RNA binding protein 2 |
Gene Aliases: |
Prbp, TRBP |
Location: |
Chr.15:102518192-102523676 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tarbp2 | NM_001253795.1 | NP_001240724.1 | ||
| NM_009319.3 | NP_033345.2 | |||
| AK143188.1 | ||||
| AK144205.1 | BAE25768.1 | |||
| AV454829.1 | ||||
| BC002028.1 | AAH02028.1 | |||
| BQ948575.1 | ||||
| U79962.1 | AAB38885.1 |

Genomic Map