Assay Details
Target Gene Details
Entrez Gene ID: | 23970 |
Gene Name: | protein kinase C and casein kinase substrate in neurons 2 |
Gene Aliases: |
AI197433 |
Location: |
Chr.15:83375607-83464606 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pacsin2 | NM_001159509.1 | NP_001152981.1 | ||
| NM_001159510.1 | NP_001152982.1 | |||
| NM_011862.3 | NP_035992.1 | |||
| AF128535.1 | AAD41780.1 | |||
| AK143828.1 | BAE25557.1 | |||
| AK154785.1 | BAE32828.1 | |||
| AK170299.1 | BAE41697.1 | |||
| AK191560.1 | ||||
| AK198672.1 | ||||
| AK198717.1 | ||||
| AK199099.1 | ||||
| AK199784.1 | ||||
| AK202155.1 | ||||
| BB860324.1 | ||||
| BC023502.1 | AAH23502.1 | |||
| BY235086.1 | ||||
| CA560112.1 | ||||
| CJ118256.1 |
More Information
Set Membership: |
|

Genomic Map