Assay Details
Target Gene Details
Entrez Gene ID: | 223881 |
Gene Name: | Rho family GTPase 1 |
Gene Aliases: |
A830014L09Rik, Arhs |
Location: |
Chr.15:98669205-98677461 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnd1 | NM_172612.3 | NP_766200.1 | ||
| AK043639.1 | BAC31604.1 | |||
| AK161870.1 | BAE36616.1 | |||
| BC048531.1 | AAH48531.1 |
More Information
Set Membership: |
|

Genomic Map