Assay Details
Target Gene Details
Entrez Gene ID: | 20649 |
Gene Name: | syntrophin, basic 1 |
Gene Aliases: |
- |
Location: |
Chr.15:55638505-55907305 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sntb1 | NM_016667.3 | 5 | 1551 | NP_057876.1 |
| U89997.1 | 5 | 1556 | AAB66697.1 |

Genomic Map