Assay Details
Target Gene Details
Entrez Gene ID: | 67959 |
Gene Name: | poly-U binding splicing factor 60 |
Gene Aliases: |
2410104I19Rik, 2810454F19Rik, SIAHBP1 |
Location: |
Chr.15:76070182-76080950 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Puf60 | NM_001164600.1 | 7 | 761 | NP_001158072.1 |
| NM_028364.2 | 8 | 842 | NP_082640.2 | |
| NM_133691.5 | 7 | 791 | NP_598452.2 | |
| AK012434.1 | 7 | 731 | ||
| AK075960.1 | 7 | 759 | ||
| AK146534.1 | 8 | 813 | ||
| AK149632.1 | 8 | 808 | BAE28998.1 | |
| AK151526.1 | 7 | 755 | BAE30474.1 | |
| AK168789.1 | 7 | 758 | BAE40622.1 | |
| AK181151.1 | 3 | 368 | ||
| AK181470.1 | ||||
| AK189376.1 | ||||
| AK189419.1 | ||||
| BC010601.1 | 7 | 761 | AAH10601.1 | |
| BC119437.1 | 5 | 498 | ||
| BC148274.1 | 5 | 498 |
More Information
Set Membership: |
|

Genomic Map