Assay Details
Target Gene Details
Entrez Gene ID: | 19013 |
Gene Name: | peroxisome proliferator activated receptor alpha |
Gene Aliases: |
4933429D07Rik, AW742785, Nr1c1, PPAR-alpha, PPARalpha, Ppar |
Location: |
Chr.15:85728192-85806851 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppara | NM_001113418.1 | 3 | 361 | NP_001106889.1 |
| NM_011144.6 | 3 | 599 | NP_035274.2 | |
| AK035676.1 | 3 | 600 | BAC29149.1 | |
| AK081709.1 | 3 | 362 | BAC38303.1 | |
| AK149460.1 | 3 | 298 | BAE28893.1 | |
| BC016892.1 | 3 | 592 | AAH16892.1 | |
| X57638.1 | 3 | 320 | CAA40856.1 |
More Information
Set Membership: |
|

Genomic Map