Assay Details
Target Gene Details
Entrez Gene ID: | 246738 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 28 |
Gene Aliases: |
BC020175, ORF28 |
Location: |
Chr.16:91614257-91618999 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc28 | NM_001099738.1 | 1 | 164 | NP_001093208.1 |
| NM_138664.2 | 1 | 164 | NP_619605.2 | |
| AK081175.1 | 1 | 160 | BAC38156.1 | |
| BB622009.1 | 1 | 164 | ||
| BC020175.1 | 1 | 158 | ||
| BU520154.1 | 1 | 169 |
More Information
Set Membership: |
|

Genomic Map