Assay Details
Target Gene Details
Entrez Gene ID: | 74663 |
Gene Name: | RIKEN cDNA 4930435E12 gene |
Gene Aliases: |
TSCPA |
Location: |
Chr.16:38812204-38828772 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930435E12Rik | NM_029042.1 | NP_083318.1 | ||
| AK015321.1 | ||||
| AK077164.1 | BAC36654.1 | |||
| BC115688.1 |
More Information
Set Membership: |
|

Genomic Map