Assay Details
Target Gene Details
Entrez Gene ID: | 12374 |
Gene Name: | calcium-sensing receptor |
Gene Aliases: |
CaR, Gprc2a |
Location: |
Chr.16:36490585-36562141 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Casr | NM_013803.2 | NP_038831.2 | ||
| AB027140.1 | BAA77688.1 | |||
| AF068900.1 | AAC19388.1 | |||
| AF110178.1 | AAD28371.1 | |||
| AF110179.1 | AAD28372.1 | |||
| AF128842.1 | AAD40638.1 | |||
| AF159565.1 | AAF00193.1 | |||
| AK029770.1 | BAC26608.1 |
More Information
Set Membership: |
|

Genomic Map