Assay Details
Target Gene Details
Entrez Gene ID: | 208211 |
Gene Name: | asparagine-linked glycosylation 1 (beta-1,4-mannosyltransferase) |
Gene Aliases: |
HMAT1, HMT1 |
Location: |
Chr.16:5233621-5244907 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alg1 | NM_145362.2 | 1 | 160 | NP_663337.2 |
| AK145871.1 | 1 | 160 | BAE26713.1 |
More Information
Set Membership: |
|

Genomic Map