Assay Details
Target Gene Details
Entrez Gene ID: | 76916 |
Gene Name: | translocase of inner mitochondrial membrane domain containing 1 |
Gene Aliases: |
2810021C21Rik, 4930455C21Rik, AV135763 |
Location: |
Chr.16:38497843-38522685 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Timmdc1 | NM_024273.2 | NP_077235.2 | ||
| AK012774.1 | BAB28461.1 | |||
| AK081781.1 | BAC38330.1 | |||
| BC002243.1 | AAH02243.1 | |||
| CJ056343.1 |

Genomic Map