Assay Details
Target Gene Details
Entrez Gene ID: | 321003 |
Gene Name: | X-prolyl aminopeptidase (aminopeptidase P) 3, putative |
Gene Aliases: |
APP3, E430012M05Rik |
Location: |
Chr.15:81400128-81457476 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xpnpep3 | NM_177310.2 | NP_796284.1 | ||
| AK088324.1 | BAC40282.1 | |||
| BC137569.1 | ||||
| BC144717.1 |
More Information
Set Membership: |
|

Genomic Map