Assay Details
Target Gene Details
Entrez Gene ID: | 76872 |
Gene Name: | coiled-coil domain containing 116 |
Gene Aliases: |
4930432J16Rik |
Location: |
Chr.16:17139064-17147146 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc116 | NM_029779.2 | NP_084055.2 | ||
| AK015290.1 | BAB29781.1 | |||
| BC050881.1 | 1 | 84 | AAH50881.1 | |
| BC057359.1 | AAH57359.1 |

Genomic Map