Assay Details
Target Gene Details
Entrez Gene ID: | 239839 |
Gene Name: | coiled-coil domain containing 14 |
Gene Aliases: |
G630039H03Rik |
Location: |
Chr.16:34690567-34725204 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc14 | NM_172824.3 | 6 | 675 | NP_766412.2 |
| AK029725.1 | 6 | 676 | ||
| AK090294.1 | 6 | 582 | BAC41158.1 | |
| AK172014.1 | 1 | 929 | ||
| BB615458.1 | ||||
| BB618371.1 | ||||
| BC031204.1 | 6 | 571 | AAH31204.1 | |
| CF742918.1 | 4 | 408 |

Genomic Map