Assay Details
Target Gene Details
Entrez Gene ID: | 13052 |
Gene Name: | coxsackie virus and adenovirus receptor |
Gene Aliases: |
2610206D03Rik, AU016810, AW553441, CAR, MCAR, MCVADR |
Location: |
Chr.16:78301460-78359785 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cxadr | NM_001025192.3 | NP_001020363.1 | ||
| NM_009988.4 | NP_034118.1 | |||
| AK004908.2 | BAB23660.2 | |||
| AK145569.1 | BAE26518.1 | |||
| AK160457.1 | BAE35798.1 | |||
| BC016457.1 | AAH16457.1 | |||
| U90715.1 | AAC53148.1 | |||
| Y10320.1 | CAA71368.1 | |||
| Y11929.1 | CAA72679.1 |

Genomic Map