Assay Details
Target Gene Details
Entrez Gene ID: | 70028 |
Gene Name: | dopey family member 2 |
Gene Aliases: |
0610038M01Rik, 2610510B01Rik, AI503246, C21orf5, mKIAA0933 |
Location: |
Chr.16:93711907-93810588 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dopey2 | NM_026700.2 | 1 | 77 | NP_080976.1 |
| NM_027293.1 | 1 | 77 | NP_081569.1 | |
| AK002808.1 | BAB22373.1 | |||
| AK147255.1 | BAE27801.1 | |||
| AK156170.1 | ||||
| AK164839.1 | BAE37938.1 | |||
| BY184348.1 | 1 | 77 | ||
| BY195296.1 | 1 | 77 |

Genomic Map