Assay Details
Target Gene Details
Entrez Gene ID: | 66793 |
Gene Name: | EF hand calcium binding domain 1 |
Gene Aliases: |
4930449A16Rik, 5430404L10Rik |
Location: |
Chr.16:14906257-14969061 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efcab1 | NM_025769.3 | 1 | 77 | NP_080045.1 |
| AK015866.1 | 1 | 98 | BAB30007.1 | |
| AK017267.1 | 1 | 78 | BAB30660.1 | |
| AK030381.1 | 1 | 87 | BAC26934.1 | |
| AK038735.1 | 1 | 87 | BAC30115.1 | |
| AK042748.1 | 1 | 119 | BAC31349.1 | |
| AK043519.1 | 1 | 49 | BAC31565.1 | |
| AK077722.1 | 1 | 80 | BAC36981.1 | |
| BC085167.1 | AAH85167.1 |

Genomic Map