Assay Details
Target Gene Details
Entrez Gene ID: | 70186 |
Gene Name: | family with sequence similarity 162, member A |
Gene Aliases: |
2310056P07Rik, HGTD-P |
Location: |
Chr.16:36043844-36071515 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam162a | NM_027342.1 | 3 | 225 | NP_081618.1 |
| AK009946.1 | 3 | 225 | BAB26601.1 | |
| BC010826.1 | 3 | 234 | AAH10826.1 |

Genomic Map