Assay Details
Target Gene Details
Entrez Gene ID: | 26425 |
Gene Name: | nucleotide binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.16:10411878-10424428 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nubp1 | NM_011955.2 | 11 | 1845 | NP_036085.1 |
| BI134234.1 | ||||
| BM938167.1 | 1 | 420 |
Target Gene Details
Entrez Gene ID: | 383103 |
Gene Name: | trans-golgi network vesicle protein 23A |
Gene Aliases: |
Fam18a |
Location: |
Chr.16:10420557-10447388 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tvp23a | NM_001013778.1 | NP_001013800.1 | ||
| BC068110.1 | AAH68110.1 |

Genomic Map