Assay Details
Target Gene Details
Entrez Gene ID: | 21819 |
Gene Name: | thyroglobulin |
Gene Aliases: |
Tgn, cog |
Location: |
Chr.15:66670756-66850721 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 25 - Intron 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tg | NM_009375.2 | NP_033401.2 | ||
| AF076186.1 | AAC32268.1 | |||
| AF076187.1 | AAC32269.1 | |||
| BC111467.1 | AAI11468.1 | |||
| U76389.1 | AAB53204.1 |
More Information
Set Membership: |
|

Genomic Map