Assay Details
Target Gene Details
Entrez Gene ID: | 14375 |
Gene Name: | X-ray repair complementing defective repair in Chinese hamster cells 6 |
Gene Aliases: |
70kDa, G22p1, Ku70 |
Location: |
Chr.15:81987826-82040085 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xrcc6 | NM_010247.2 | NP_034377.2 | ||
| AB010282.1 | BAA28874.1 | |||
| AF483500.1 | AAL90774.1 | |||
| AF483501.1 | AAL90775.1 | |||
| AK143570.1 | BAE25441.1 | |||
| AK146392.1 | BAE27135.1 | |||
| AK146394.1 | BAE27137.1 | |||
| AK146396.1 | BAE27139.1 | |||
| AK178816.1 | ||||
| BC031422.1 | AAH31422.1 | |||
| M38700.1 | AAA39396.1 |
More Information
Set Membership: |
|

Genomic Map