Assay Details
Target Gene Details
Entrez Gene ID: | 108105 |
Gene Name: | UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5 |
Gene Aliases: |
beta3GnT5 |
Location: |
Chr.16:19760192-19772758 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3gnt5 | NM_001159407.1 | NP_001152879.1 | ||
| NM_001159408.1 | NP_001152880.1 | |||
| NM_054052.3 | NP_473393.2 | |||
| AK032911.1 | BAC28082.1 | |||
| AK037166.1 | ||||
| AK048010.1 | BAC33213.1 | |||
| AK083839.1 | BAC39037.1 | |||
| AY029203.1 | AAK31579.1 | |||
| BC063076.1 | AAH63076.1 | |||
| CN533455.1 |
More Information
Set Membership: |
|

Genomic Map