Assay Details
Target Gene Details
Entrez Gene ID: | 224044 |
Gene Name: | cytochrome P450, family 2, subfamily ab, polypeptide 1 |
Gene Aliases: |
EG224044, F730023N20 |
Location: |
Chr.16:20308387-20325412 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2ab1 | NM_183158.3 | NP_898981.2 | ||
| AK089408.1 | BAC40871.1 | |||
| BC138088.1 |
More Information
Set Membership: |
|

Genomic Map