Assay Details
Target Gene Details
Entrez Gene ID: | 224405 |
Gene Name: | cysteine and tyrosine-rich protein 1 |
Gene Aliases: |
- |
Location: |
Chr.16:85450357-85550905 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyyr1 | NM_144853.3 | NP_659102.1 | ||
| AF442733.1 | AAL35295.1 | |||
| AK031840.1 | ||||
| AY061854.1 | AAL35738.1 | |||
| BC099957.1 | AAH99957.1 | |||
| BC125594.1 | ||||
| BC125596.1 |

Genomic Map