Assay Details
Target Gene Details
Entrez Gene ID: | 70028 |
Gene Name: | dopey family member 2 |
Gene Aliases: |
0610038M01Rik, 2610510B01Rik, AI503246, C21orf5, mKIAA0933 |
Location: |
Chr.16:93711907-93810588 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dopey2 | NM_027293.1 | NP_081569.1 | ||
| AK122403.1 | BAC65685.1 | |||
| AK147255.1 | BAE27801.1 | |||
| AK147536.1 | BAE27979.1 | |||
| BC057081.1 | AAH57081.1 |

Genomic Map