Assay Details
Target Gene Details
Entrez Gene ID: | 70967 |
Gene Name: | eva-1 homolog C (C. elegans) |
Gene Aliases: |
1700092M14Rik, 4931408A02Rik, Fam176c |
Location: |
Chr.16:90826719-90904885 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eva1c | NM_001199210.1 | NP_001186139.1 | ||
| NM_027627.2 | NP_081903.1 | |||
| AF358257.1 | AAL40410.1 | |||
| AK016443.1 | BAB30238.1 | |||
| AK037486.1 | BAC29817.1 | |||
| BC100350.1 | AAI00351.1 | |||
| BC132305.1 | ||||
| BC157969.1 |

Genomic Map