Assay Details
Target Gene Details
Entrez Gene ID: | 22631 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta polypeptide |
Gene Aliases: |
1110013I11Rik, 14-3-3zeta, AI596267, AL022924, AU020854 |
Location: |
Chr.15:36770261-36802984 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ywhaz | NM_001253805.1 | NP_001240734.1 | ||
| NM_001253806.1 | NP_001240735.1 | |||
| NM_001253807.1 | NP_001240736.1 | |||
| NM_011740.3 | NP_035870.1 | |||
| AK083368.1 | BAC38887.1 | |||
| AK145657.1 | BAE26570.1 | |||
| AK146800.1 | BAE27442.1 | |||
| AK150381.1 | BAE29512.1 | |||
| AK151900.1 | BAE30783.1 | |||
| AK162099.1 | BAE36724.1 | |||
| AK167128.1 | BAE39275.1 | |||
| AK187117.1 | ||||
| AK201135.1 | ||||
| BC050891.1 | AAH50891.1 | |||
| BC089334.1 | AAH89334.1 | |||
| CF172373.1 | ||||
| D78647.1 | BAA11464.1 | |||
| D83037.1 | BAA11751.1 | |||
| D87660.1 | BAA13421.1 | |||
| U79231.1 | AAC53254.1 |
More Information
Set Membership: |
|

Genomic Map