Assay Details
Target Gene Details
Entrez Gene ID: | 56375 |
Gene Name: | UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 4 |
Gene Aliases: |
9130402O08Rik, B4galt-IV, b4Gal-T4, beta4Gal-T4, beta4GalT-IV |
Location: |
Chr.16:38742103-38769054 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4galt4 | NM_001285793.1 | NP_001272722.1 | ||
| NM_019804.4 | NP_062778.2 | |||
| AF142672.1 | AAF22222.1 | |||
| AF158746.1 | AAF80363.1 | |||
| AK018658.1 | ||||
| AK041692.1 | BAC31035.1 | |||
| AK045602.1 | BAC32433.1 | |||
| AK050005.1 | BAC34028.1 | |||
| AK052013.1 | BAC34832.1 | |||
| AK053603.1 | BAC35443.1 | |||
| AK085368.1 | BAC39433.1 | |||
| AK136042.1 | ||||
| AK136637.1 | BAE23084.1 | |||
| AK214305.1 | ||||
| BC013492.1 | AAH13492.1 | |||
| BC031115.1 | AAH31115.1 |
More Information
Set Membership: |
|

Genomic Map