Assay Details
Target Gene Details
Entrez Gene ID: | 223642 |
Gene Name: | zinc finger CCCH type containing 3 |
Gene Aliases: |
BC049953, Kiaa0150, Smicl, Zc3hdc3 |
Location: |
Chr.15:75754434-75841971 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zc3h3 | NM_172121.1 | NP_742119.1 | ||
| AJ516034.1 | CAD56773.1 | |||
| AK138281.1 | BAE23611.1 | |||
| BC049953.1 | AAH49953.1 | |||
| BC060682.1 | AAH60682.1 | |||
| BC111448.1 |

Genomic Map