Assay Details
Target Gene Details
Entrez Gene ID: | 209200 |
Gene Name: | deltex 3-like (Drosophila) |
Gene Aliases: |
AU042200, BC023741 |
Location: |
Chr.16:35926515-35939027 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dtx3l | NM_001013371.2 | 4 | 2212 | NP_001013389.2 |
| AK146802.1 | 4 | 2120 | BAE27443.1 | |
| BC137694.1 | 4 | 2153 |
More Information
Set Membership: |
|

Genomic Map